2,12-Dichlorocyclododecanone ≥98.0% (通过GC)

供应商: TCI EUROPE
Warning

TCIAD2911-1GEA 0 CNY
TCIAD2911-1G
2,12-Dichlorocyclododecanone ≥98.0% (通过GC)
2,12-Dichlorocyclododecanone
分子式: C₁₂H₂₀Cl₂O
分子量: 251,19 g/mol
存储条件: Refrigerator
MDL编号: MFCD04117917
CAS号: 858830-26-9

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR