2-(2,6-Dichlorophenyl)ethanol ≥97.0% (通过GC)

供应商: TCI EUROPE

TCIAD2922-5GEA 0 CNY
TCIAD2922-5G
2-(2,6-Dichlorophenyl)ethanol ≥97.0% (通过GC)
2-(2,6-Dichlorophenyl)ethanol
分子式: C₈H₈Cl₂O
分子量: 191,05 g/mol
沸点: 152°C
MDL编号: MFCD00800674
CAS号: 30595-79-0

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Melting point 78.0 to 83.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR