2-(二甲基乙烯基硅)吡啶 ≥97.0% (通过GC)

供应商: TCI EUROPE

Synonyms: 二甲基(2-吡啶基)乙烯基硅烷

TCIAD2935-1GEA 0 CNY
TCIAD2935-1G
2-(二甲基乙烯基硅)吡啶 ≥97.0% (通过GC)
2-(二甲基乙烯基硅)吡啶
分子式: C₉H₁₃NSi
分子量: 163,30 g/mol
存储条件: Refrigerator
MDL编号: MFCD06797092
CAS号: 321155-39-9

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR