6-[[7-(N,N-Dimethylaminosulphonyl)-2,1,3-benzoxadiazol-4-yl]amino]hexanoic acid ≥98.0% (通过HPLC,滴定分析)
供应商: TCI EUROPE
TCIAD3178-100MGEA
0
CNY
TCIAD3178-100MG
6-[[7-(N,N-Dimethylaminosulphonyl)-2,1,3-benzoxadiazol-4-yl]amino]hexanoic acid ≥98.0% (通过HPLC,滴定分析)
6-[[7-(N,N-Dimethylaminosulphonyl)-2,1,3-benzoxadiazol-4-yl]amino]hexanoic acid
分子式:
C₁₄H₂₀N₄O₅S 分子量: 356,40 g/mol |
CAS号:
1820741-40-9 |
产品技术规格
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 152.0 to 156.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...