2,2'-Dithiobis(6-fluorobenzoic acid) ≥98.0% (通过HPLC,滴定分析)
供应商: TCI EUROPE
|
Warning
|
TCIAD3198-5GEA
0
CNY
TCIAD3198-5G
2,2'-Dithiobis(6-fluorobenzoic acid) ≥98.0% (通过HPLC,滴定分析)
2,2'-Dithiobis(6-fluorobenzoic acid)
分子式:
C₁₄H₈F₂O₄S₂ 分子量: 342,33 g/mol 溶点: 265°C |
CAS号:
147027-64-3 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...