3',6'-Dimethoxyfluoran ≥98.0% (通过HPLC)

供应商: TCI EUROPE

TCIAD3205-25GEA 1125 CNY
TCIAD3205-25G
3',6'-Dimethoxyfluoran ≥98.0% (通过HPLC)
3',6'-Dimethoxyfluoran
分子式: C₂₂H₁₆O₅
CAS号: 36886-76-7
EINECS: 253-255-7

现在订购

产品技术规格

Appearance White to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 199.0 to 205.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR