2,4-Diamino-6-[2-(2-undecyl-1-imidazolyl)ethyl]-1,3,5-triazine ≥99.0% (通过滴定分析)
供应商: TCI EUROPE
|
Warning
|
TCIAD3239-25GEA
0
CNY
TCIAD3239-25G
2,4-Diamino-6-[2-(2-undecyl-1-imidazolyl)ethyl]-1,3,5-triazine ≥99.0% (通过滴定分析)
2,4-Diamino-6-[2-(2-undecyl-1-imidazolyl)ethyl]-1,3,5-triazine
分子式:
C₁₉H₃₃N₇ 分子量: 359,52 g/mol |
MDL编号:
MFCD00047352 CAS号: 50729-75-4 EINECS: 256-742-2 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 99.0 % |
Melting point | 191.0 to 195.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...