6-硝基-2,3-二羟基喹喔啉 ≥95.0% (通过HPLC)
供应商: TCI EUROPE
|
Warning
|
Synonyms:
6-硝基喹喔啉-2,3-二酮
TCIAD3241-5GEA
0
CNY
TCIAD3241-5G
6-硝基-2,3-二羟基喹喔啉 ≥95.0% (通过HPLC)
6-硝基-2,3-二羟基喹喔啉
分子式:
C₈H₅N₃O₄ 分子量: 207,15 g/mol |
CAS号:
2379-56-8 |
产品技术规格
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...