4,5-Dicyano-1,3-dithiol-2-one ≥98.0% (通过GC)

供应商: TCI EUROPE
Danger

TCIAD3252-1GEA 1925 CNY
TCIAD3252-1G
4,5-Dicyano-1,3-dithiol-2-one ≥98.0% (通过GC)
4,5-Dicyano-1,3-dithiol-2-one
分子式: C₅N₂OS₂
CAS号: 934-31-6
UN号: 3439
ADR: 6.1,III

现在订购

产品技术规格

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 122.0 to 126.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR