1,4-Dibutoxybenzene ≥98.0% (通过GC)

供应商: TCI EUROPE

TCIAD3264-5GEA 0 CNY
TCIAD3264-5G
1,4-Dibutoxybenzene ≥98.0% (通过GC)
1,4-Dibutoxybenzene
分子式: C₁₄H₂₂O₂
分子量: 222,33 g/mol
沸点: 158°C
CAS号: 104-36-9
EINECS: 203-196-8

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 44.0 to 48.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR