4,4'-(1,3-Dimethylbutylidene)diphenol ≥98.0% (通过GC)

供应商: TCI EUROPE
Danger

TCIAD3267-25GEA 330 CNY
TCIAD3267-25G
4,4'-(1,3-Dimethylbutylidene)diphenol ≥98.0% (通过GC)
4,4'-(1,3-Dimethylbutylidene)diphenol
分子式: C₁₈H₂₂O₂
分子量: 270,37 g/mol
CAS号: 6807-17-6
EINECS: 401-720-1
UN号: 3077
ADR: 9,III

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 153.0 to 157.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR