9,9-Dimethylfluorene-2-carboxylic acid ≥98.0% (通过GC,滴定分析)
供应商: TCI EUROPE
TCIAD3332-1GEA
1275
CNY
TCIAD3332-1G
9,9-Dimethylfluorene-2-carboxylic acid ≥98.0% (通过GC,滴定分析)
9,9-Dimethylfluorene-2-carboxylic acid
分子式:
C₁₆H₁₄O₂ |
CAS号:
28320-62-9 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 194.0 to 198.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...