3,5-Dinitro-L-tyrosine sodium salt ≥98.0% (通过HPLC)
供应商: TCI EUROPE
TCIAD3379-1GEA
825
CNY
TCIAD3379-1G
3,5-Dinitro-L-tyrosine sodium salt ≥98.0% (通过HPLC)
3,5-Dinitro-L-tyrosine sodium salt
分子式:
C₉H₉N₃O₇ 分子量: 293,17 g/mol |
CAS号:
502481-30-3 |
产品技术规格
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | 97.0 to 103.0% (calcd.on anh.substance) |
Specific rotation [a]20/D | +9.5 to +11.5 deg (C=1, 1mol/L HCl) |
Water | max. 5.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...