5,10-二氢-5,10-二甲基吩嗪 ≥99.0% (通过GC)

供应商: TCI EUROPE
Warning

TCIAD3392-1GEA 0 CNY
TCIAD3392-1G
5,10-二氢-5,10-二甲基吩嗪 ≥99.0% (通过GC)
5,10-二氢-5,10-二甲基吩嗪
分子式: C₁₄H₁₄N₂
分子量: 210,28 g/mol
MDL编号: MFCD00092904
CAS号: 15546-75-5

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (GC) min. 99.0 %
Melting point 152.0 to 155.0 deg-C
Solubility in Toluene almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR