6,11-二氢二苯并[b,e]硫杂卓-11-酮 ≥98.0% (通过GC)
供应商: TCI EUROPE
|
Warning
|
TCIAD3416-5GEA
0
CNY
TCIAD3416-5G
TCIAD3416-25G
6,11-二氢二苯并[b,e]硫杂卓-11-酮 ≥98.0% (通过GC)
6,11-二氢二苯并[b,e]硫杂卓-11-酮
分子式:
C₁₄H₁₀OS 分子量: 226,29 g/mol 沸点: 180°C |
MDL编号:
MFCD02093738 CAS号: 1531-77-7 EINECS: 216-241-1 |
产品技术规格
Appearance | White to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 85.0 to 89.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...