3,4-Diethoxyphenylacetic acid ≥98.0% (通过GC,滴定分析)

供应商: TCI EUROPE
Warning

TCIAD3419-5GEA 565 CNY
TCIAD3419-5G
3,4-Diethoxyphenylacetic acid ≥98.0% (通过GC,滴定分析)
3,4-Diethoxyphenylacetic acid
分子式: C₁₂H₁₆O₄
分子量: 224,26 g/mol
MDL编号: MFCD00040785
CAS号: 38464-04-9
EINECS: 253-957-3

现在订购

产品技术规格

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 78.0 to 82.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR