2-(3,5-Dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97.0% (通过GC)
供应商: TCI EUROPE
|
Warning
|
TCIAD3533-5GEA
620
CNY
TCIAD3533-5G
2-(3,5-Dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97.0% (通过GC)
2-(3,5-Dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
分子式:
C₁₄H₂₁BO₂ 分子量: 232,13 g/mol |
CAS号:
325142-93-6 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...