1-(2,4-Dichlorophenyl)-2-(1-imidazolyl)ethanol ≥98.0% (通过GC,滴定分析)

供应商: TCI EUROPE

TCIAD3629-25GEA 0 CNY
TCIAD3629-25G
1-(2,4-Dichlorophenyl)-2-(1-imidazolyl)ethanol ≥98.0% (通过GC,滴定分析)
1-(2,4-Dichlorophenyl)-2-(1-imidazolyl)ethanol
分子式: C₁₁H₁₀Cl₂N₂O
分子量: 257,11 g/mol
MDL编号: MFCD00044708
CAS号: 24155-42-8
EINECS: 246-042-5

现在订购

产品技术规格

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 133.0 to 139.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR