Diethyl-2-chloro-3-oxosuccinate ≥93.0% (通过GC)

供应商: TCI EUROPE

TCIAD3635-1GEA 0 CNY
TCIAD3635-1G
Diethyl-2-chloro-3-oxosuccinate ≥93.0% (通过GC)
Diethyl-2-chloro-3-oxosuccinate
分子式: C₈H₁₁ClO₅
分子量: 222,6229 g/mol
沸点: 130°C
存储条件: Refrigerator
MDL编号: MFCD04114359
CAS号: 34034-87-2

现在订购

产品技术规格

Appearance White to Yellow to Green clear liquid
Purity (GC) min. 93.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR