3,4-Dihydroxy-5-nitrobenzaldehyde ≥98.0% (通过GC)

供应商: TCI EUROPE
Danger

TCIAD3645-25GEA 0 CNY
TCIAD3645-25G
3,4-Dihydroxy-5-nitrobenzaldehyde ≥98.0% (通过GC)
3,4-Dihydroxy-5-nitrobenzaldehyde
分子式: C₇H₅NO₅
分子量: 183,12 g/mol
MDL编号: MFCD00871897
CAS号: 116313-85-0
EINECS: 441-810-8

现在订购

产品技术规格

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 95.0 %
Melting point 145.0 to 149.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR