10,11-Dihydro-11-oxodibenzo[b,f][1,4]thiazepine ≥98.0% (通过HPLC,总氮)

供应商: TCI EUROPE

TCIAD3693-25GEA 0 CNY
TCIAD3693-25G
10,11-Dihydro-11-oxodibenzo[b,f][1,4]thiazepine ≥98.0% (通过HPLC,总氮)
10,11-Dihydro-11-oxodibenzo[b,f][1,4]thiazepine
分子式: C₁₃H₉NOS
分子量: 227,2817 g/mol
溶点: 265°C
存储条件: Refrigerator
MDL编号: MFCD00901197
CAS号: 3159-07-7

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR