1,3-Dimethoxy-1,1,3,3-tetraphenyldisiloxane ≥90.0% (通过GC)

供应商: TCI EUROPE
Warning

TCIAD3734-5GEA 0 CNY
TCIAD3734-5G
1,3-Dimethoxy-1,1,3,3-tetraphenyldisiloxane ≥90.0% (通过GC)
1,3-Dimethoxy-1,1,3,3-tetraphenyldisiloxane
分子式: C₂₆H₂₆O₃Si₂
分子量: 442,66 g/mol
MDL编号: MFCD05663939
CAS号: 94593-08-5

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 90.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR