2,2-Dimethyl-5-(3-thienyl)-1,3-dioxane-4,6-dione ≥98.0% (通过HPLC)
供应商: TCI EUROPE
TCIAD3759-25GEA
0
CNY
TCIAD3759-25G
2,2-Dimethyl-5-(3-thienyl)-1,3-dioxane-4,6-dione ≥98.0% (通过HPLC)
2,2-Dimethyl-5-(3-thienyl)-1,3-dioxane-4,6-dione
分子式:
C₁₀H₁₀O₄S 分子量: 226,25 g/mol 溶点: 130°C |
MDL编号:
MFCD08458279 CAS号: 61857-83-8 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...