2,5-Dimethyl-3(2H)-furanone ≥98.0% (通过GC)

供应商: TCI EUROPE
Warning

TCIAD3795-5GEA 0 CNY
TCIAD3795-5G
2,5-Dimethyl-3(2H)-furanone ≥98.0% (通过GC)
2,5-Dimethyl-3(2H)-furanone
分子式: C₆H₈O₂
分子量: 112,13 g/mol
沸点: 72°C
闪点: 52°C
MDL编号: MFCD00052571
CAS号: 14400-67-0
EINECS: 238-365-5
UN号: 1993
ADR: 3,III

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR