1,3-Dihydro-1-methoxyisobenzofuran ≥97.0% (通过GC)
供应商: TCI EUROPE
TCIAD3828-5GEA
0
CNY
TCIAD3828-5G
1,3-Dihydro-1-methoxyisobenzofuran ≥97.0% (通过GC)
1,3-Dihydro-1-methoxyisobenzofuran
分子式:
C₉H₁₀O₂ 分子量: 150,18 g/mol 沸点: 106°C |
CAS号:
67536-29-2 |
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...