2,2'-Dibromo-9,9'-spirobi[9H-fluorene] ≥95.0% (通过GC)

供应商: TCI EUROPE
Warning

TCIAD3872-1GEA 0 CNY
TCIAD3872-1G
2,2'-Dibromo-9,9'-spirobi[9H-fluorene] ≥95.0% (通过GC)
2,2'-Dibromo-9,9'-spirobi[9H-fluorene]
分子式: C₂₅H₁₄Br₂
分子量: 474,20 g/mol
溶点: 240°C
MDL编号: MFCD08704219
CAS号: 67665-47-8

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR