1-Docosanethiol ≥98.0% (通过滴定分析)

供应商: TCI EUROPE
Warning

TCIAD3905-5GEA 0 CNY
TCIAD3905-5G
1-Docosanethiol ≥98.0% (通过滴定分析)
1-Docosanethiol
分子式: C₂₂H₄₆S
分子量: 342,67 g/mol
MDL编号: MFCD02262163
CAS号: 7773-83-3

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Purity (Iodometric Titration) min. 98.0 %
Melting point 46.0 to 50.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR