9,10-Dihydro-9,10-bis(2-carboxyethyl)-N-(4-nitrophenyl)-10,9-(epoxyimino)anthracene-12-carboxamide
供应商: TCI EUROPE
|
Warning
|
TCIAD3929-100MGEA
0
CNY
TCIAD3929-100MG
9,10-Dihydro-9,10-bis(2-carboxyethyl)-N-(4-nitrophenyl)-10,9-(epoxyimino)anthracene-12-carboxamide
9,10-Dihydro-9,10-bis(2-carboxyethyl)-N-(4-nitrophenyl)-10,9-(epoxyimino)anthracene-12-carboxamide
分子式:
C₂₇H₂₃N₃O₈ 存储条件: Refrigerator |
CAS号:
1225023-87-9 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...