4,7-Dibromo-2,1,3-benzoselenadiazole ≥98.0% (通过GC)

供应商: TCI EUROPE
Danger

TCIAD3944-1GEA 1475 CNY
TCIAD3944-1G
4,7-Dibromo-2,1,3-benzoselenadiazole ≥98.0% (通过GC)
4,7-Dibromo-2,1,3-benzoselenadiazole
分子式: C₆H₂Br₂N₂Se
溶点: 287°C
CAS号: 63224-42-0
UN号: 3283
ADR: 6.1,III

现在订购

产品技术规格

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR