1,8-二氮双环[5.4.0]十一烯溴酸盐 ≥98.0% (通过HPLC,总氮)
供应商: TCI EUROPE
|
Danger
|
TCIAD3976-5GEA
0
CNY
TCIAD3976-5G
1,8-二氮双环[5.4.0]十一烯溴酸盐 ≥98.0% (通过HPLC,总氮)
1,8-二氮双环[5.4.0]十一烯溴酸盐
分子式:
C₉H₁₆N₂ 分子量: 392,96 g/mol |
MDL编号:
MFCD00191982 CAS号: 138666-59-8 UN号: 3261 ADR: 8,II |
产品技术规格
Appearance | Light yellow to Amber to Dark green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 121.0 to 125.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...