3,5-二甲基-4-硝基吡啶-2-甲醇 ≥98.0% (通过GC,滴定分析)
供应商: TCI EUROPE
|
Warning
|
TCIAD4026-5GEA
0
CNY
TCIAD4026-5G
3,5-二甲基-4-硝基吡啶-2-甲醇 ≥98.0% (通过GC,滴定分析)
3,5-二甲基-4-硝基吡啶-2-甲醇
分子式:
C₈H₁₀N₂O₃ 分子量: 182,18 g/mol |
MDL编号:
MFCD03426035 CAS号: 149082-03-1 |
产品技术规格
Appearance | White to Gray to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 66.0 to 70.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...