Barium disodium ethylenediaminetetraacetate hydrate ≥98.0% (通过滴定分析)

供应商: TCI EUROPE
Danger

TCIAE0085-25GEA 0 CNY
TCIAE0085-25G
Barium disodium ethylenediaminetetraacetate hydrate ≥98.0% (通过滴定分析)
Barium disodium ethylenediaminetetraacetate hydrate
分子式: C₁₀H₁₆N₂O₈
分子量: 471,52 g/mol
MDL编号: MFCD00054444
CAS号: 18744-58-6
UN号: 1564
ADR: 6.1,III

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Chelometric Titration) min. 98.0 % (calcd.on anh.substance)
Solubility in Water almost transparency
Water 12.0 to 18.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR