2-Ethylhexylamine hydrochloride ≥97.0% (通过总氮和滴定分析)

供应商: TCI EUROPE

TCIAE0128-25GEA 0 CNY
TCIAE0128-25G
2-Ethylhexylamine hydrochloride ≥97.0% (通过总氮和滴定分析)
2-Ethylhexylamine hydrochloride
分子式: C₈H₁₉N
分子量: 165,71 g/mol
MDL编号: MFCD00060201
CAS号: 26392-49-4
EINECS: 247-648-2

现在订购

产品技术规格

Appearance White to Almost white wax
Purity (with Total Nitrogen) min. 97.0 %
Purity (Argentmetric Titration) min. 97.0 %
Solubility in Water very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR