4'-Ethoxybenzylidene-4-cyanoaniline ≥98.0% (通过GC,总氮)

供应商: TCI EUROPE
Warning

TCIAE0240-1GEA 0 CNY
TCIAE0240-1G
4'-Ethoxybenzylidene-4-cyanoaniline ≥98.0% (通过GC,总氮)
4'-Ethoxybenzylidene-4-cyanoaniline
分子式: C₁₆H₁₄N₂O
分子量: 250,30 g/mol
MDL编号: MFCD00043561
CAS号: 24742-30-1
EINECS: 246-445-6

现在订购

产品技术规格

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (with Total Nitrogen) min. 98.0 %
Mesomorphic range 101.0 to 123.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR