3-(乙基氨基)-4-甲酚 ≥92.0% (通过GC)
供应商: TCI EUROPE
|
Warning
|
TCIAE0252-25GEA
0
CNY
TCIAE0252-25G
3-(乙基氨基)-4-甲酚 ≥92.0% (通过GC)
3-(乙基氨基)-4-甲酚
分子式:
C₉H₁₃NO 分子量: 151,21 g/mol |
MDL编号:
MFCD00002382 CAS号: 120-37-6 EINECS: 204-391-0 |
产品技术规格
Appearance | White to Gray to Red powder to crystal |
Purity (GC) | min. 92.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...