3-Ethyl-3-methylglutarimide ≥98.0% (通过GC)
供应商: TCI EUROPE
|
Danger
|
TCIAE0284-10GEA
0
CNY
TCIAE0284-10G
3-Ethyl-3-methylglutarimide ≥98.0% (通过GC)
3-Ethyl-3-methylglutarimide
分子式:
C₈H₁₃NO₂ 分子量: 155,20 g/mol |
MDL编号:
MFCD00006673 CAS号: 64-65-3 EINECS: 200-588-0 UN号: 2811 ADR: 6.1,II |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 125.0 to 129.0 deg-C |
Solubility in Methanol | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...