3-乙基-3-甲基戊二酸 ≥98.0% (通过滴定分析)

供应商: TCI EUROPE
Warning

Synonyms: 3-乙基-甲基戊烷二酸

TCIAE0285-5GEA 715 CNY
TCIAE0285-5G
3-乙基-3-甲基戊二酸 ≥98.0% (通过滴定分析)
3-乙基-3-甲基戊二酸
分子式: C₈H₁₄O₄
分子量: 174,20 g/mol
MDL编号: MFCD00020494
CAS号: 5345-01-7
EINECS: 226-294-2

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 82.0 to 86.0 deg-C
Solubility in Water very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR