3-Ethyl-3-methylheptane ≥99.0% (通过GC)

供应商: TCI EUROPE
Danger

TCIAE0304-1MLEA 0 CNY
TCIAE0304-1ML
3-Ethyl-3-methylheptane ≥99.0% (通过GC)
3-Ethyl-3-methylheptane
分子式: C₁₀H₂₂
分子量: 142,29 g/mol
MDL编号: MFCD00048812
CAS号: 17302-01-1
UN号: 3295
ADR: 3,III

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR