1-Ethyl-3-hydroxypiperidine ≥98.0% (通过GC,滴定分析)

供应商: TCI EUROPE
Warning

TCIAE0376-25MLEA 0 CNY
TCIAE0376-25ML
1-Ethyl-3-hydroxypiperidine ≥98.0% (通过GC,滴定分析)
1-Ethyl-3-hydroxypiperidine
分子式: C₇H₁₅NO
分子量: 129,20 g/mol
闪点: 47°C
MDL编号: MFCD00006508
CAS号: 13444-24-1
EINECS: 236-584-0
UN号: 1993
ADR: 3,III

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR