1-Ethyl-3-guanidinothiourea hydrochloride ≥98.0% (通过滴定分析)
供应商: TCI EUROPE
TCIAE0419-5GEA
0
CNY
TCIAE0419-5G
1-Ethyl-3-guanidinothiourea hydrochloride ≥98.0% (通过滴定分析)
1-Ethyl-3-guanidinothiourea hydrochloride
分子式:
C₄H₁₀N₄S 分子量: 182,67 g/mol |
MDL编号:
MFCD00191430 CAS号: 381670-29-7 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 157.0 to 160.0 deg-C |
Solubility in Water | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...