N-乙基-3-吡咯烷醇 ≥95.0% (通过GC,滴定分析)

供应商: TCI EUROPE
Warning

TCIAE0425-10MLEA 0 CNY
TCIAE0425-10ML
N-乙基-3-吡咯烷醇 ≥95.0% (通过GC,滴定分析)
N-乙基-3-吡咯烷醇
分子式: C₆H₁₃NO
分子量: 115,18 g/mol
沸点: 110°C
闪点: 73°C
MDL编号: MFCD00003179
CAS号: 30727-14-1
EINECS: 250-310-7

现在订购

产品技术规格

Appearance Colorless to Brown clear liquid
Purity (GC) min. 95.0 %
Purity (Nonaqueous Titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR