(3R)-(+)-3-(Ethylamino)pyrrolidine ≥97.0% (通过GC,滴定分析)

供应商: TCI EUROPE
Danger

TCIAE0433-1GEA 2050 CNY
TCIAE0433-1G
(3R)-(+)-3-(Ethylamino)pyrrolidine ≥97.0% (通过GC,滴定分析)
(3R)-(+)-3-(Ethylamino)pyrrolidine
分子式: C₆H₁₄N₂
分子量: 114,19 g/mol
沸点: 176°C
MDL编号: MFCD00191358
CAS号: 381670-30-0
UN号: 2734
ADR: 8,II

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %
Specific rotation [a]20/D +12.0 to +17.0 deg (C=10, EtOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR