3-Ethyl-2-methyl-3-pentanol ≥98.0% (通过GC)

供应商: TCI EUROPE
Warning

TCIAE0481-5GEA 0 CNY
TCIAE0481-5G
3-Ethyl-2-methyl-3-pentanol ≥98.0% (通过GC)
3-Ethyl-2-methyl-3-pentanol
分子式: C₈H₁₈O
分子量: 130,23 g/mol
沸点: 59°C
MDL编号: MFCD00046589
CAS号: 597-05-7
UN号: 1987
ADR: 3,III

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR