9-Ethylcarbazole-3-carboxaldehyde-N-ethyl-N-(o-tolyl)hydrazone ≥98.0% (通过HPLC)

供应商: TCI EUROPE

TCIAE0577-1GEA 0 CNY
TCIAE0577-1G
9-Ethylcarbazole-3-carboxaldehyde-N-ethyl-N-(o-tolyl)hydrazone ≥98.0% (通过HPLC)
9-Ethylcarbazole-3-carboxaldehyde-N-ethyl-N-(o-tolyl)hydrazone
分子式: C₂₄H₂₅N₃
分子量: 355,49 g/mol
MDL编号: MFCD06797102
CAS号: 1274948-12-7

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Solubility in Toluene very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR