Ethyl benzo[d]isoxazole-3-carboxylate ≥98.0% (通过GC)

供应商: TCI EUROPE

TCIAE0731-100MGEA 0 CNY
TCIAE0731-100MG
Ethyl benzo[d]isoxazole-3-carboxylate ≥98.0% (通过GC)
Ethyl benzo[d]isoxazole-3-carboxylate
分子式: C₁₀H₉NO₃
分子量: 191,1834 g/mol
MDL编号: MFCD06659787
CAS号: 57764-49-5

现在订购

产品技术规格

Purity (GC) min. 98.0 %
Melting point 57.0 to 61.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR