4-Ethynyltriphenylamine ≥98.0% (通过HPLC,总氮)

供应商: TCI EUROPE
Warning

TCIAE0894-1GEA 0 CNY
TCIAE0894-1G
4-Ethynyltriphenylamine ≥98.0% (通过HPLC,总氮)
4-Ethynyltriphenylamine
分子式: C₂₀H₁₅N
分子量: 269,35 g/mol
MDL编号: MFCD09030769
CAS号: 205877-26-5

现在订购

产品技术规格

Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 109.0 to 113.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR