α-雌二醇 ≥98.0% (通过GC)

供应商: TCI EUROPE
Warning

Synonyms: 雌二醇1,3,5(10)-雌甾三烯-3,17α-二醇3,17α-二羟基-1,3,5(10)-雌三烯

TCIAE0919-1GEA 0 CNY
TCIAE0919-1G
α-雌二醇 ≥98.0% (通过GC)
α-雌二醇
分子式: C₁₈H₂₄O₂
分子量: 272,39 g/mol
MDL编号: MFCD00064144
CAS号: 57-91-0
EINECS: 200-354-8

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 222.0 to 226.0 deg-C
Specific rotation [a]20/D +46.0 to +52.0 deg (C=1, Dioxane)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR