2-Methylpentyl formate ≥90.0% (通过GC)
供应商: TCI EUROPE
|
Warning
|
TCIAF0174-5MLEA
0
CNY
TCIAF0174-5ML
2-Methylpentyl formate ≥90.0% (通过GC)
2-Methylpentyl formate
分子式:
C₇H₁₄O₂ 分子量: 130,19 g/mol 沸点: 148°C |
MDL编号:
MFCD00059817 CAS号: 381670-34-4 UN号: 3272 ADR: 3,III |
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 90.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...