6-Fluoro-2-methyl-4-chromanone ≥97.0% (通过GC)
供应商: TCI EUROPE
TCIAF0271-5GEA
0
CNY
TCIAF0271-5G
6-Fluoro-2-methyl-4-chromanone ≥97.0% (通过GC)
6-Fluoro-2-methyl-4-chromanone
分子式:
C₁₀H₉FO₂ |
MDL编号:
MFCD00059072 CAS号: 88754-96-5 |
产品技术规格
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 69.0 to 72.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...