1-(甲酰氨基甲基)-1H-苯并三唑 ≥98.0% (通过HPLC)

供应商: TCI EUROPE
Warning

TCIAF0339-5GEA 0 CNY
TCIAF0339-5G
1-(甲酰氨基甲基)-1H-苯并三唑 ≥98.0% (通过HPLC)
1-(甲酰氨基甲基)-1H-苯并三唑
分子式: C₈H₈N₄O
分子量: 176,18 g/mol
MDL编号: MFCD00134328
CAS号: 87022-36-4

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 137.0 to 141.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR